Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ninjurin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572821 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572821 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572821 Supplier Invitrogen™ Supplier No. PA572821
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ninjurin 1 Polyclonal Antibody for ELISA, IHC (P)

Homophilic cell adhesion molecule that promotes axonal growth. May play a role in nerve regeneration and in the formation and function of other tissues. Cell adhesion requires divalent cations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ninjurin 1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Ninj1
Gene Accession No. O70131, P70617, Q92982
Gene Alias AU024536; Nerve injury-induced protein 1; nerve injury-induced protein-1; NIN1; Ninj1; NINJURIN; ninjurin 1; ninjurin-1
Gene Symbols Ninj1
Host Species Rabbit
Immunogen NINJ1 antibody for a 15 amino acid peptide near the carboxy terminus of human NINJ1. The immunogen is within the last 50 amino acids of NINJ1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18081, 25338, 4814
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.