Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niobium n-propoxide, 99% (metals basis), Thermo Scientific™

Catalog No. AA3657106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3657106 5 g
AA3657114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3657106 Supplier Thermo Scientific Chemicals Supplier No. 3657106

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38874-17-8
Molecular Formula C15H40NbO5
Molecular Weight (g/mol) 393.386
MDL Number MFCD00144917
InChI Key VXIXBNJIHXAVQS-UHFFFAOYSA-N
Synonym niobium n-propoxide
PubChem CID 131878811
IUPAC Name niobium;propan-1-ol
SMILES CCCO.CCCO.CCCO.CCCO.CCCO.[Nb]

Specifications

Boiling Point 166°C (0.05 mmHg)
Linear Formula Nb(OC3H7)5
Quantity 5 g
UN Number UN1993
Sensitivity Moisture sensitive
Formula Weight 388.35
Percent Purity 99%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Niobium n-propoxide
Hazard Category H226-H315-H319
Hazard Statement GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.