Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niobium(V) oxide, Puratronic™, 99.9985% (metals basis)

Catalog No. AA1082230
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1082230 250 g
AA1082218 50 g
AA10822A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1082230 Supplier Thermo Scientific Chemicals Supplier No. 01082230
Add to Cart

CAS: 1313-96-8 | Nb2O5 | 265.81 g/mol

Niobium pentoxide is the precursor to a majority of niobium compounds, powder metallurgy products, and alloys. It is also used in glass, optic and ceramic applications. The nanoparticles of niobium pentoxide are useful in the preparation of electrochemical capacitors and cathodes. In addition, appropriate preparations of this compound can be used as a dietary supplement in food industry. It is a precursor to lithium niobate which finds extensive use in piezoelectric sensors, anti-aliasing filters, optical waveguides, mobile phones, surface acoustic devices, optical modulators, including linear and non-linear optical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-96-8
Molecular Formula Nb2O5
Molecular Weight (g/mol) 265.81
MDL Number MFCD00011128
InChI Key ZKATWMILCYLAPD-UHFFFAOYSA-N
Synonym niobium oxide, niobium v oxide, niobium pentoxide, diniobium pentaoxide, niobia, niobium pentaoxide, diniobium pentoxide, niobium 5+ oxide, nobum oxde
PubChem CID 123105
SMILES O=[Nb](=O)O[Nb](=O)=O

Specifications

Melting Point 1520°C
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.9985% (metals basis)
Solubility Information Insoluble in water; soluble in hydrogen fluoride.
IUPAC Name dioxoniobiooxy(dioxo)niobium
Formula Weight 265.81
Odor Odorless
Appearance White powder or lump
Density 4.6
Chemical Name or Material Niobium(V) oxide
Show More Show Less
Hazard Category H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.
EINECSNumber 215-213-6
RTECSNumber QU0500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.