Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NIPA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570648
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570648 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570648 Supplier Invitrogen™ Supplier No. PA570648
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%.

This gene encodes a magnesium transporter that associates with early endosomes and the cell surface in a variety of neuronal and epithelial cells. This protein may play a role in nervous system development and maintenance. Multiple transcript variants encoding different isoforms have been found for this gene. Mutations in this gene have been associated with autosomal dominant spastic paraplegia 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NIPA1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NIPA1
Gene Accession No. Q7RTP0
Gene Alias 1110027G09Rik; A830014A18Rik; FSP3; Magnesium transporter NIPA1; NIPA magnesium transporter 1; NIPA1; non imprinted in Prader-Willi/Angelman syndrome 1; non imprinted in Prader-Willi/Angelman syndrome 1 homolog; non imprinted in Prader-Willi/Angelman syndrome 1 homolog (human); non-imprinted in Prader-Willi/Angelman syndrome 1; non-imprinted in Prader-Willi/Angelman syndrome region protein 1; Non-imprinted in Prader-Willi/Angelman syndrome region protein 1 homolog; spastic paraplegia 6 (autosomal dominant); spastic paraplegia 6 homolog; spastic paraplegia 6 protein; Spg6
Gene Symbols NIPA1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human NIPA1 (aa 280-329).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123606
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.