Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NIPA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567168
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567168 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567168 Supplier Invitrogen™ Supplier No. PA567168
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LESLPVSFRKD EKAMNGNLSN MYEVLNNNEE S Highest antigen sequence identity to the following orthologs - mouse 88%, rat 88%.

Nipa2 encodes a protein that acts as a selective Mg(2+) transporter. The Nipa2 gene encodes a possible magnesium transporter, and is located adjacent to the imprinted domain in the Prader-Willi syndrome deletion region of chromosome 15. Alternate splicing of the Nipa2 gene results in multiple transcript variants. Pseudogenes of the Nipa2 gene are found on chromosomes 3, 7 and 21. Diseases associated with NIPA2 include Angelman Syndrome and Prader-Willi Syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NIPA2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NIPA2
Gene Accession No. Q8N8Q9
Gene Alias 2600017P10Rik; 3830408P04Rik; AB041581; Magnesium transporter NIPA2; MNCb-2146; Nipa2; non imprinted in Prader-Willi/Angelman syndrome 2; non imprinted in Prader-Willi/Angelman syndrome 2 homolog; non imprinted in Prader-Willi/Angelman syndrome 2 homolog (human); non-imprinted in Prader-Willi/Angelman syndrome region protein 2; Non-imprinted in Prader-Willi/Angelman syndrome region protein 2 homolog; RGD1306051
Gene Symbols NIPA2
Host Species Rabbit
Immunogen Recombinant Human NIPA2. Recombinant protein control fragment (Product #RP-105532).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81614
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.