Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NIPBL Antibody [mFluor Violet 500 SE], Novus Biologicals™
SDP

Catalog No. NB798861
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB798861 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB798861 Supplier R&D Systems Supplier No. NB10093320MFV500

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

NIPBL Polyclonal antibody specifically detects NIPBL in Human samples. It is validated for Immunoprecipitation,Western Blot

Specifications

Antigen NIPBL
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias CDLS1, delangin, DKFZp434L1319, FLJ11203, FLJ12597, FLJ13354, FLJ13648, FLJ44854, IDN3-B, IDN3CDLS, Nipped-B homolog (Drosophila), nipped-B-like protein, Scc2, SCC2 homolog, sister chromatid cohesion 2 homolog
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1025 and 1075 of human Nipped-B-like using the numbering given in entry NP_597677.2 (GeneID 25836).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 25836
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.