Learn More
TOKU-E NISIN SOLN 1ML
Supplier:  TOKU-E N0841ML
Encompass_Preferred
Antibiotic, ReadyMade™, sterile-filtered, 20,000-40,000 IU/ml in 0.02N HCl, Useful in COVID-19 research, cell culture, food safety
Catalog No. NC2068748
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.