Learn More
Apexbio Technology LLC Lonidamine 50264-69-2 10mM (in 1mL DMSO)
Supplier: Apexbio Technology LLC B19645.1
Lonidamine (AF-1890) is a cell metabolism inhibitor targeting mitochondrial pyruvate carrier and hexokinase exhibiting inhibitory activity with a Ki value of approximately 2 5 M Through these targets it disrupts glycolytic metabolism by impairing aerobic glycolysis processes common in tumor cells Researchers utilize Lonidamine primarily to investigate metabolic pathways implicated in cancer cell energy production alterations in mitochondrial functions and related metabolic disorders Additionally Lonidamine is employed experimentally in studies examining mitochondrial dysfunction-associated conditions and inflammatory responses including pulmonary fibrosis
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.