Learn More
Beckman Coulter NIST Traceable Particle Size Controls
Particle Size Controls for use with LS 13 320 XR Analyzers to verify instrument and module performance.
Supplier:  Beckman Coulter C12539
 View more versions of this product
Catalog No. 15-655-203
Only null left
Add to Cart
Description
Description
  • For verifying instrument and module performance
  • Recommended for LS 13 320 XR instruments
  • NIST traceable
Specifications
Specifications
Particle Size Control
Polystyrene
Verifying Instrument and Module Perfomance
15 mL
0.30 μm
LS 13 320 XR Analyzer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.