Learn More
Beckman Coulter NIST Traceable Particle Size Controls
Particle Size Controls for use with LS 13 320 XR Analyzers to verify instrument and module performance.
Supplier: Beckman Coulter C18128
Description
- For verifying instrument and module performance
- Recommended for LS 13 320 XR instruments
- NIST traceable
Specifications
Particle Size Control | |
0.085 μm, 0.20 μm, 0.50 μm | |
LS 13 320 XR Analyzer |
Polystyrene | |
Verifying Instrument and Module Perfomance |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.