Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nital Etchant 10% Nitric Acid in Ethanol, Reagents

Nital Etchant 10% Nitric Acid in Ethanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Reagents Holdings Llc CN2015004A

Quantity 4 L

 View more versions of this product

Catalog No. 01-334-612


Only null left
Add to Cart

Description

Description

Specifications

Chemical Identifiers

64-17-5 , 7697-37-2 , 7732-18-5
46.07
Nitric Acid in Ethyl Alcohol
CCO
C2H6O
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol

Specifications

Colorless to Light Yellow
76°C
Nitric Acid in Ethyl Alcohol
CCO
46.07
Reagent
0.8703
4 L
Liquid
64-17-5 , 7697-37-2 , 7732-18-5
C2H6O
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
10%
Poly Bottle
<7
Non-sterile
Nital Etchant 10% Nitric Acid in Ethanol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Accessories
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.