Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nital Etchant, 3% Nitric Acid in Methanol, Reagents

Catalog No. 01334620
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-620 2.5 L
01-334-621 1 L
01-334-619 250 mL
01-334-618 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-334-620 Supplier Reagents Holdings Llc Supplier No. CN2012002.5D1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nital Etchant, 3% Nitric Acid in Methanol Meets ASTM E407 Specification Etchant 74, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-56-1 , 7697-37-2 , 7732-18-5
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Nitric Acid in Methyl Alcohol
IUPAC Name methanol
SMILES CO

Specifications

Color Colorless
Boiling Point 71°C
Percent Purity 3%
Grade Reagent
Packaging Glass Bottle
Specific Gravity 0.8081
pH <1
Quantity 2.5 L
Sterility Non-sterile
Physical Form Liquid
Chemical Name or Material Nital Etchant, 3% Nitric Acid in Methanol Meets ASTM E407 Specification Etchant 74
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.