Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nital Etchant, 3% Nitric Acid in Methanol, Reagents

Nital Etchant, 3% Nitric Acid in Methanol Meets ASTM E407 Specification Etchant 74, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

$53.90 - $170.00

Chemical Identifiers

CAS 67-56-1 , 7697-37-2 , 7732-18-5
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Nitric Acid in Methyl Alcohol
IUPAC Name methanol
SMILES CO
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
01-334-621
Accessories View Documents
Reagents Holdings Llc
CN2012001C
1 L
Each for $93.80
Add to Cart
 
01-334-620
Accessories View Documents
Reagents Holdings Llc
CN2012002.5D1
2.5 L
Each for $170.00
Add to Cart
 
01-334-619
Accessories View Documents
Reagents Holdings Llc
CN201200250C
250 mL
Each for $53.90
Add to Cart
 
01-334-618
Accessories View Documents
Reagents Holdings Llc
CN201200500C
500 mL
Each for $69.75
Add to Cart
 
Description

Description

Specifications

Chemical Identifiers

67-56-1 , 7697-37-2 , 7732-18-5
32.04
Nitric Acid in Methyl Alcohol
CO
CH4O
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol

Specifications

Colorless
71°C
Nitric Acid in Methyl Alcohol
CO
32.04
Reagent
0.8081
Non-sterile
Nital Etchant, 3% Nitric Acid in Methanol Meets ASTM E407 Specification Etchant 74
67-56-1 , 7697-37-2 , 7732-18-5
CH4O
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
3%
Glass Bottle
<1
Liquid
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.