Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nital Etchant, 5% (v/v) in Alcohol, Ricca Chemical
SDP

Catalog No. R53183001C Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber Glass Bottle
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R53183001C 1 L Amber Glass Bottle
R531830025D 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R53183001C Supplier Ricca Chemical Company Supplier No. R53183001C
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Nital Etchant, 5% (v/v) in Alcohol
Color Colorless
CAS 64-17-5, 7697-37-2, 67-56-1, 7732-18-5
CAS Min % 86.57, 6.38, 4.33, 2.73
CAS Max % 86.57, 6.38, 4.33, 2.73
Density 0.825
Solubility Information Miscible in water
Percent Purity 5%
Grade Laboratory
Odor Alcoholic
Packaging Amber Glass Bottle
pH Acidic
Quantity 1 L
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.