Learn More
AdipoGen Nitazoxanide (250 mg)
SDP
Supplier:  AdipoGen AGCR13723M250
Encompass_Preferred
Chemical. CAS: 55981-09-4. Formula: C12H9N3O5S. MW: 307.3. Nitazoxanide is a broad-spectrum antiparasitic, antimicrobial and antiviral drug that is used in medicine for the treatment of various helminthic, protozoal, bacterial and viral infections. Nitazoxanide is rapidly metabolized to tizaxonide, an antiparasitic drug of the thiazolide class. The anti-protozoal activity and activity against anaerobic bacteria is believed to be due to interference with the pyruvate:ferredoxin oxidoreductase (PFOR) enzyme-dependent electron transfer reaction which is essential to anaerobic energy metabolism. It is being studied for potential treatment for chronic hepatitis B, chronic hepatitis C, rotavirus, norovirus gastroenteritis and the coronavirus SARS-CoV-2 (COVID-19). The mechanism is suppressing the viral replication by inhibiting maturation of the viral hemagglutinin and the viral transcription factor immediate early 2 (IE2).
Catalog No. 50-205-4126
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.