Learn More
Lkt Laboratories CAS 104206-65-7, ≥99%, HPPD inhibitor. It is used to slow the effects of hereditary type 1 tyrosinemia.
SDP
Supplier:  Lkt Laboratories N3476
Encompass_Preferred
Nitisinone is a nitrobenzene inhibitor of HPPD, an enzyme involved in tyrosine degradation. Nitisinone is clinically used to slow the effects of hereditary type 1 tyrosinemia and as a potential treatment for alkaptonuria.
Catalog No. NC2522205
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.