Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrate Nitrogen Standard, 1000 ppm N (4427 ppm NO3-), Ricca Chemical
SDP

Catalog No. 54594 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
5459-4 120 mL Natural Poly Bottle
5459-16 500 mL Natural Poly Bottle
13-622-612 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 5459-4 Supplier Ricca Chemical Company Supplier No. 54594
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7757-79-1 , 7732-18-5
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
PubChem CID 24434
ChEBI CHEBI:63043
IUPAC Name potassium nitrate
SMILES [K+].[O-][N+]([O-])=O

Specifications

Packaging Natural Poly Bottle
CAS Min % 97.0, 0.7
CAS Max % 100.0, 0.73
Methods ASTM/EPA for Nitrate Nitrogen
Equivalence 1mL = 1mgN
Boiling Point 100°C
Quantity 120 mL
Grade Not Available
Concentration or Composition (by Analyte or Components) 1000ppm N as nitrate, 4427ppm NO3-
Chemical Name or Material Nitrogen Standard (as Nitrate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.