Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrate Standard, 1000 ppm NO3- (226 ppm N), Ricca Chemical
SDP

Catalog No. 53074 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
16 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5307-4 120 mL
5307-16 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 5307-4 Supplier Ricca Chemical Company Supplier No. 53074
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7757-79-1 , 7732-18-5
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
PubChem CID 24434
ChEBI CHEBI:63043
IUPAC Name potassium nitrate
SMILES [K+].[O-][N+]([O-])=O

Specifications

Packaging Natural Poly Bottle
CAS Min % 98.0, 0.16
CAS Max % 100.0, 0.17
Boiling Point 100°C
Quantity 120 mL
Chemical Name or Material Nitrate Standard, 1 mL = 1 mg NO3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.