Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrendipine, Tocris Bioscience™
SDP

Catalog No. 060150 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-015-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 06-015-0 Supplier R&D Systems Supplier No. 0601/50

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CaV1.x blocker

L-type calcium channel blocker. Induces autophagy.

Chemical Identifiers

CAS 39562-70-4
Molecular Formula C18H20N2O6
Molecular Weight (g/mol) 360.366
InChI Key PVHUJELLJLJGLN-UHFFFAOYSA-N
Synonym nitrendipine, bayotensin, baypress, nidrel, nitrendipinum, nitrendipino, bylotensin, nitrendipin, deiten, nitrepin
PubChem CID 4507
ChEBI CHEBI:7582
IUPAC Name 5-O-ethyl 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(NC(=C(C1C2=CC(=CC=C2)[N+](=O)[O-])C(=O)OC)C)C

Specifications

Quantity 50 mg
Formula Weight 360.37
Percent Purity >99%
Chemical Name or Material Nitrendipine
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.