Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety NitriShield™ 6010 Nitrile Disposable Gloves

Catalog No. 19812737 Shop All MCR Safety Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Color:
Green
Orange
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
19-812-737 Large Green
19-812-738 Medium Green
19-812-740 Large Orange
19-812-741 Medium Orange
19-812-743 X-Large Orange
19-812-739 X-Large Green
19-812-742 Small Orange
19-812-744 2X-Large Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-812-737 Supplier MCR Safety Supplier No. 6010GL
This item is not returnable. View return policy

NitriShield™ medical grade nitrile gloves are used in applications that require resistance to punctures, tears or exposure to certain types of chemicals. Available in green or orange.

  • Green or orange nitrile
  • Medical Grade, Class 1
  • Powder free
  • Rolled cuff
  • Latex-free
  • Resistant to punctures and tears
  • Textured grip

Specifications

Certifications/Compliance Class 1 Medical Device
Certifications/Compliance (Europe) Class B according the EN374:2016 standard
Chemotherapeutic Rated No
Applications Healthcare, Laboratory
Color Green
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 5.1 mil
Finger Thickness (Metric) 0.12 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 4.3 mil
Palm Thickness (Metric) 0.10 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Large
Sterility Non-sterile
Thickness (English) 4 mil
Thickness (Metric) 0.10 mm
FDA 510 (k) Yes
Glove Cuff Style Rolled
Model 6010
Series NitriShield™
Length (English) 9.5 in.
Length (Metric) 24.13 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.