Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Nitri-Task KL™ Nitrile Coated Aramid Fiber Gloves

Catalog No. 191500996
Change view
Click to view available options
Size:
Small
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-150-0996 X-Large
19-150-0993 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-150-0996 Supplier Honeywell™ North™ Supplier No. NFKL13/10XL
This item is not returnable. View return policy

Lightweight shell is constructed from a blend of Lycra and aramid fibers for protection against cuts and abrasion. Textured nitrile coating on the palm and fingers enhances grip capabilities.

  • Kevlar™ aramid fiber provides a high level of cut protection, as well as impermeability to oils
  • Knit construction conforms snugly to the hand for dexterity and tactility
  • Nitrile-coating enhances grip in a wide variety of environments
  • Dark color is soil-proof for an extended glove life
  • Ventilated back for optimized breathability
TRUSTED_SUSTAINABILITY

Specifications

Coating Material Nitrile
Color Black
Exterior Finish Fully Textured
Hand Specific
Material Aramid Fiber, Lycra
Product Type Seamless Knit Gloves
Size X-Large
For Use With (Application) Automotive Assembly, Metal Handling, and General Industry
Glove Cuff Color Natural
Glove Cuff Style Knit
Series NFKL13
Yarn Gauge 13
Length (English) 9.8 in.
Length (Metric) 25 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.