Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, 1:1, 50% v/v, Reagents

Catalog No. 01334634
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
Safety Coated Glass
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-334-634 1 L Safety Coated Glass
01-334-635 1 L Glass Bottle
01-334-633 2.5 L Glass Bottle
01-334-632 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-334-634 Supplier Reagents Holdings Llc Supplier No. CN2003001C1
Add to Cart
Add to Cart

Nitric Acid, 1:1, 50% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Azotic Acid, Hydrogen nitrate
IUPAC Name water
SMILES O

Specifications

pH <1
Color Colorless to Light Yellow
Boiling Point 107°C
Packaging Safety Coated Glass
Specific Gravity 1.2629
Quantity 1 L
Physical Form Liquid
Percent Purity 50%
Grade Reagent
Chemical Name or Material Nitric Acid, 1:1, 50% v/v
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.