Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, 10% (v/v), Ricca Chemical
SDP

Catalog No. 53205 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
20 L
Packaging:
Cubitainer
Hard Pack
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
53-205 20 L Cubitainer
5320-16 500 mL Natural Poly Bottle
5320-32 1 L Natural Poly Bottle
R532000020E 20 L Hard Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 53-205 Supplier Ricca Chemical Company Supplier No. 53205
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Appearance of Solution Colorless to Light Yellow Liquid
Packaging Cubitainer
Quantity 20 L
Synonym hydrogen nitrate
Percent Purity 10% V/V
Grade Laboratory
Chemical Name or Material Nitric Acid
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.