Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, 30% (v/v) in Methanol, Ricca Chemical
SDP

Catalog No. R53203001C1 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Natural Poly Bottle
Safety-coated Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R53203001C1 1 L Safety-coated Glass Bottle
R53203001A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R53203001C1 Supplier Ricca Chemical Company Supplier No. R53203001C1
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67-56-1 , 7697-37-2 , 7732-18-5
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
IUPAC Name methanol
SMILES CO

Specifications

Appearance of Solution Colorless Liquid
Concentration 29.8 to 30.2 N
CAS Max % 56.49, 30.45, 13.05
Packaging Safety-coated Glass Bottle
Quantity 1 L
Percent Purity 30% V/V
Grade Laboratory
Chemical Name or Material Nitric Acid in Methanol
ShelfLife 12 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.