Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, 5% (v/v) in Denatured Ethanol, Ricca Chemical
SDP

Catalog No. 5318132 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
5318-132 1 L Natural Poly Bottle
5318-15 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5318-132 Supplier Ricca Chemical Company Supplier No. 5318.132
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64-17-5 , 7697-37-2 , 67-56-1 , 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Colorless to Yellow
CAS Min % 4.0, 90.0, 4.0
CAS Max % 6.0, 91.0, 6.0
Packaging Natural Poly Bottle
Quantity 1 L
Physical Form Liquid
Percent Purity 5% V/V
Grade Laboratory
Concentration or Composition (by Analyte or Components) 5% (v/v) Alcoholic Solution
Chemical Name or Material Nitric Acid
ShelfLife 12 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.