Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric acid, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., for determinations with dithizone, ≥65%, Solstice

Catalog No. 6001105 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
Packaging:
Glass Bottle
Plastic Bottle
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-011-05 2.5 L Plastic Bottle
60-045-841 4 x 2.5 L Glass Bottle
60-045-840 4 x 2.5 L Poly Bottle
60-011-04 1 L Glass Bottle
60-011-03 1 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-011-05 Supplier Solstice Supplier No. 307092.5L

Puriss. p.a., Reag. ISO, Reag. Ph. Eur., for determinations with dithizone,≥65%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Density 1.37 to 1.41g/mL at 20°C
Packaging Plastic Bottle
Residue after Ignition ≤0.0005% (as SO4)
Vapor Pressure 8mmHg (20°C)
Quantity 2.5 L
UN Number 2031
Formula Weight 63.01g/mol
Percent Purity ≥65%
Grade puriss. p.a.
Chemical Name or Material Nitric Acid, Reag. Ph. Eur. reag. ISO
Hazard Statement P210-P220-P260-P280-P305 + P351 + P338-P370 + P378
Precautionary Statement H272-H290-H314
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.