Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, Technical, Spectrum™ Chemical
SDP

Catalog No. 18604839 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
6 x 500 mL
4 x 2.5 L
200 L
Packaging:
Fiber Drum
Glass Bottle
Plastic Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-839 6 x 500 mL Glass Bottle
18-604-838 500 mL Glass Bottle
18-604-835 2.5 L Glass Bottle
18-604-834 2.5 L Plastic Bottle
18-604-837 4 x 2.5 L Glass Bottle
18-604-840 200 L Fiber Drum
18-604-836 4 x 2.5 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-604-839 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. N1088500MLGLCS6
Add to Cart
Add to Cart

N1088, 7697-37-2, HNO3

Spectrum™ Chemical Nitric Acid, Technical is a strong oxidizing agent and a highly corrosive mineral acid. Primarily used as a reagent, the pure compound is colorless and used for nitration, typically to an organic molecule.

Chemical Identifiers

CAS 7697-37-2 , 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

CAS Max % 70%, 30%
Packaging Glass Bottle
Quantity 6 x 500 mL
UN Number UN2031
Grade Technical
Chemical Name or Material Nitric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.