Learn More
Abcam Nitric Oxide Assay Kit, Cell-based, Flow Cytometry, 100 Tests
Description
- Assay type: Cell-based
- Detection method: Flow cytometry-fluorescent
- Sample types: Suspension cells, Adherent cells
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| No. of Reactions | 100 Tests |
| For Use With (Equipment) | Flow Cytometer |
| Format | Kit |
| Detection Location | In-Cell Detection |
| Target Molecule | Nitric Oxide |
| Detection Method | Fluorescence |
| Sample Type | Suspension cells, Adherent cells |
| Quantity | 100 Tests |
| For Use With (Application) | Flow Cytometry |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.