Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Nitrile Coated Seamless Knit Nylon Gloves

Catalog No. 19050283D Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
7
8
9
10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-050-283D 10
19-050-283A 7
19-050-283B 8
19-050-283C 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-050-283D Supplier SHOWA™ Supplier No. 450010
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Flat dipped nitrile-coated gloves provide abrasion resistance and enhanced grip capabilities in dry conditions.

  • Breathable, ventilated back
  • Seamless nylon liner for increased comfort
  • Knit wrist prevents dirt and debris from entering the glove
  • Machine washable

Specifications

Certifications/Compliance EN 388:2016+A1:2018 4121XX
Certifications/Compliance (Europe) CE, EN 388:2016+A1:2018 4121XX
Coated Area Palm, Fingers
Construction Seamless Knit
Cuff Material Elastic
Coating Material Nitrile
Color Green
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size 10
EN388 Standard 4121XX
For Use With (Application) Engineering, Component Assembly, Automotive Repairs, and Parts Handling
Gender Unisex
Glove Cuff Color Natural
Glove Cuff Style Knit
Grip Design Smooth
Liner Material Nylon
Packaging Type Polybag
Series 4500
Length (English) 10 in.
Length (Metric) 25.4 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.