Learn More
Description
- Recycling solution for nitrile gloves
- All brands of nitrile gloves are accepted
- Eco-friendly Boxes: Made from recycled materials and easy to assemble
- Collection boxes include a self-locking mechanisms and prepaid shipping label
- Not for recycling of non-nitrile gloves, including latex, neoprene, butyl, vinyl and glove blends
- Do not recycle biological materials, infectious waste, bloodborne pathogens, hazardous materials or gloves exposed to more than 10% chemical exposure of the surface area
- Available in 3 sizes: Small 12 x 12 x 20 in., Medium 12 x 11 x 38 in., Large 15 x 13 x 38 in.
- 5 boxes per case
- Quarterly report available upon request: Reports to include, but not limited to, materials recycled, amount processed, and materials end-of-life scenarios.
Specifications
Specifications
| Capacity (Metric) | 82.20 L |
| Capacity (English) | 18.08 gal. |
| Dimensions (L x W x H) | 12 x 11 x 38 in. |
| Size | Medium |
| Autoclavable | Not Autoclavable |
| For Use With (Equipment) | Nitrile Gloves |
| Material | Recycled Materials |
| Product Type | Recycling Box |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
