Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrite (as Nitrogen) Standard, 1000ppm, Preserved with Chloroform (1mL = 1mg N, 1mL = 3.28mg NO2-), Certified, LabChem™

Catalog No. LC179301 Shop All LabChem Products
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC179301 500 mL Poly Bottle
LC179307 125 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC179301 Supplier LabChem, Inc. Supplier No. LC179301
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7758-09-0 , 67-66-3 , 64-17-5 , 7732-18-5
Molecular Formula KNO2
Molecular Weight (g/mol) 85.10
MDL Number MFCD00011408
InChI Key BXNHTSHTPBPRFX-UHFFFAOYSA-M
PubChem CID 516910
IUPAC Name potassium nitrite
SMILES [K+].[O-]N=O

Specifications

Packaging Poly Bottle
CAS Max % 99.19, 0.61, 0.198, 0.002
Identification Passes Test
Density 1g/mL
Physical Form Liquid
Concentration 1000 ppm ±10 ppm (1mL = 1mg N, 1mL = 3.28mg NO2- )
Linear Formula KNO2
Color Colorless
Solubility Soluble in water
Quantity 500 mL
Formula Weight 85.1
Grade Certified
Assay as N
Chemical Name or Material Nitrite (as Nitrogen) Standard, 1000ppm, Preserved with Chloroform (1mL = 1mg N, 1mL = 3.28mg NO2-)
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Suspected of causing cancer.
Harmful to aquatic life.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Avoid release to the environment.
Wear protective gloves, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Refrigerate
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.