Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrite Standard, 1000 μg/mL, ISO 17034, 125 mL

Catalog No. USICC007
Encompass
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USICC007 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USICC007 Supplier Agilent Technologies Supplier No. ICC007
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7632-00-0 , 7732-18-5
Molecular Formula NNaO2
Molecular Weight (g/mol) 69.00
MDL Number MFCD00011118
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
PubChem CID 23668193
ChEBI CHEBI:78870
IUPAC Name sodium nitrite
SMILES [Na+].[O-]N=O

Specifications

CAS Max % 99.9, 0.1
Analyte or Component Names Water,nitrite,soluble compounds as NO2
Element Nitrite
Solvent or Matrix Water
Quantity 125 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material Nitrite Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.