Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrite Standard, 1000 ppm NO2- (304.5 ppm N), Ricca Chemical
SDP

Catalog No. 544516 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
5445-16 500 mL Natural Poly Bottle
5445-4 120 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5445-16 Supplier Ricca Chemical Company Supplier No. 544516
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5

Specifications

Packaging Natural Poly Bottle
Equivalence 1mL = 1mg NO2 (1000ppm NO2),304.5ppm Nitrogen (N)
Boiling Point 100°C
Quantity 500 mL
Chemical Name or Material Nitrite Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.