Learn More
Ansell Bioclean™ Bouffant Cap
Catalog No. 19223709 Shop All Ansell Products
Change view
Click to view available options
Size:
One Size
Catalog No. Size
19-223-709 One Size
1 options
This item is not returnable. View return policy
Catalog No. 19-223-709 Supplier Ansell Supplier No. BBCW
Add to Cart
Add to Cart
This item is not returnable. View return policy
Lightweight and breathable bouffant reduces the risk of contamination from the head area in controlled environments.
  • Universal Size
  • Lightweight, soft non-woven Polypropylene
  • Hypoallergenic
  • Compatible with a variety of environments including cleanrooms
  • Manufactured in a facility holding ISO 9001:2008 Certifications
  • In accordance with ISO 2859
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Bouffant
Size One Size
Material Spunbond Polypropylene
Color White
Disposable Single-use
Model BBC-W
Seam Type Elasticized Double Stitching
Certifications/Compliance ISO 9001:2008 Certifications in accordance with ISO 2859
Cleanroom Class Class 10 (ISO 4)
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.