Learn More
Ansell™ BioClean™ Clearview™ Autoclavable Goggles
BIOPHARMA
Catalog No. 19668190 Shop All Ansell Products
Change view
Click to view available options
Lens Coating:
Scratch-resistant
Catalog No. Lens Coating
19-668-190 Scratch-resistant
1 options
This item is not returnable. View return policy
Catalog No. 19-668-190 Supplier Ansell™ Supplier No. BCAG
  • $2,400.00
    $1,374.65 / Dozen of 12
    Save $1,025.35
    43% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $10,424.00
    $6,279.65 / Case of 5 DZ
    Save $4,144.35
    40% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Constructed in super-soft thermoplastic rubber to provide unrivalled comfort, BioClean Clearview™ Autoclavable Cleanroom Goggles feature upper vents, indirect lower vents, and have a toughened anti-scratch polycarbonate lens. Provided non-sterile. To be autoclaved at 121°C/250°F for 30 minutes
  • Non-Sterile and Sealed in a PE Bag to maintain cleanliness
  • Upper & Indirect Ventilation holes
  • EN 166:2001 Optical Class 1
  • Toughened Anti-Scratch Polycarbonate lens
  • Tested to over 40 Autoclave Cycles
  • Super-soft thermoplastic provides unrivaled comfort
  • Processed and packed in our NEBB Certified ISO Class 4 (Class 10) Cleanroom
  • Replacement lens available (BCAG-SL 19-668-191)
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant
Certifications/Compliance ISO 9001:2015, (EU)2016/425, EN 166:2001
Product Type Autoclavable Goggles
No. per Case 12/Cs.
No. per Pack 1 Ea.
Ventilation Details Indirect base ventilation
Material Polycarbonate