Learn More
Ansell BioClean-D™ Disposable Long Sleeve Cover
Catalog No. 19668341 Shop All Ansell Products
Change view
Click to view available options
Sterility:
Non-sterile
Catalog No. Sterility
19-668-341 Non-sterile
1 options
Catalog No. 19-668-341 Supplier Ansell Supplier No. BDSCL
Add to Cart
Add to Cart
Extra long length sleeve covers are constructed from antistatic, low-linting CleanTough™ material and are ISO Class 4 compatible.
  • Protective sleeves available in sterile or non-sterile
  • Low linting, anti-static CleanTough™ fabric
  • Elasticated arms and cuffs
  • ISO Class 4 and EU GMP Grade A compatible
  • Universal size
  • Overall length: 500 mm
  • Applications include cleanroom, pharma, controlled environments, 503B facilities/compounding, manufacturing, industrial
TRUSTED_SUSTAINABILITY
Specifications
Product Type Controlled Environment Sleeve
Size Standard
Material Double Laminate Microporous Fabric
Sterility Non-sterile
Disposable Single-use
Closure Type Elastic Ends
Model BDSC
For Use With (Application) Cleanroom, Clinical, Medical, Laboratory
Certifications/Compliance ASTM D6978-05, ASTM F739-12, ISO 9001:2015
Certifications/Compliance (Europe) (EU)2016/425, EN ISO 13034:2005+A1:2009, EN ISO 14325, EN 1149-5:2008, EN 530, EN 7854, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2
Length (English) 19.6 in.
Length (Metric) 500 mm
Packaging 6 Bags, 30/Bag
Cleanroom Class Class 10 (ISO 4)
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.