Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitro Blue Tetrazolium, Fisher BioReagents

Catalog No. BP1081 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP108-1 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BP108-1

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in conjunction with BCIP as a substrate for acid and alkaline phosphatases

Nitro BT is used in conjunction with BCIP as a substrate for acid and alkaline phosphatases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-83-9
Molecular Formula C40H30Cl2N10O6
Molecular Weight (g/mol) 817.644
InChI Key FSVCQIDHPKZJSO-UHFFFAOYSA-L
Synonym NBT, Nitro BT, p-Nitro Blue Tetrazolium
PubChem CID 9281
ChEBI CHEBI:9505
IUPAC Name 2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride
SMILES COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-]

Specifications

Melting Point 189°C
Extinction Coefficient 650g(-1) cm(-1) min.
Identification Pass Test
Lambda Max 258 to 262nm
Formula Weight 817.65
Packaging Amber Glass
Color Yellow
Quantity 1 g
Physical Form Powder
Chemical Name or Material Nitro Blue Tetrazolium
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Harmful if swallowed. Avoid contact with skin and eyes. Do not breathe dust. Do not breathe vapors or spray mist. Do not ingest. Use explosion-proof equipment. Use only non-sparking tools. Use personal protective equipment. Wash off immediately with soap and plenty of water removing all contaminated clothes and shoes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Remove from exposure, lie down.

NFPA

Health:2
Flammability:1
Instability:0
Recommended Storage 4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.