CAS RN 298-83-9
Thermo Scientific Chemicals Nitro Blue Tetrazolium chloride, tech. 90%
CAS: 298-83-9 Molecular Formula: C40H30Cl2N10O6 Molecular Weight (g/mol): 817.644 MDL Number: MFCD00150013 InChI Key: FSVCQIDHPKZJSO-UHFFFAOYSA-L Synonym: nitrotetrazolium blue chloride,nitro bt,nitro blue tetrazolium,nbt,nitro blue tetrazolium chloride,p-nitrotetrazolium blue,nitrotetrazolium blue,nitroblue tetrazolium chloride,p-nitro blue tetrazolium chloride,nitro tetrazolium bt PubChem CID: 9281 ChEBI: CHEBI:9505 IUPAC Name: 2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride SMILES: COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-]
Thermo Scientific Chemicals Nitro Blue Tetrazolium Chloride, 90%
CAS: 298-83-9 Molecular Formula: C40H30Cl2N10O6 Molecular Weight (g/mol): 817.65 InChI Key: FSVCQIDHPKZJSO-UHFFFAOYSA-L Synonym: nitrotetrazolium blue chloride,nitro bt,nitro blue tetrazolium,nbt,nitro blue tetrazolium chloride,p-nitrotetrazolium blue,nitrotetrazolium blue,nitroblue tetrazolium chloride,p-nitro blue tetrazolium chloride,nitro tetrazolium bt PubChem CID: 9281 ChEBI: CHEBI:9505 IUPAC Name: 2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride SMILES: COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-]
Thermo Scientific Chemicals Nitro Blue Tetrazolium Chloride monohydrate, 90%
CAS: 298-83-9 | C40H30Cl2N10O6 | 817.644 g/mol
p-Nitro blue tetrazolium chloride, MP Biomedicals™
CAS: 298-83-9 Molecular Formula: C40H30Cl2N10O6 Molecular Weight (g/mol): 817.644 MDL Number: MFCD00012159 InChI Key: FSVCQIDHPKZJSO-UHFFFAOYSA-L Synonym: 3,3'-(3,3'-Dimethoxy-4,4'-biphenylene)-bis-(2-p-nitrophenyl)-5-(phenyl)-2H-tetrazolium chloride,NBT PubChem CID: 9281 ChEBI: CHEBI:9505 IUPAC Name: 2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride SMILES: COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-]
MilliporeSigma™ Calbiochem™ p-Nitroblue Tetrazolium Chloride
NADPH-diaphorase substrate that competitively inhibits nitric oxide synthase (IC50 = 3-4 μM).
p-Nitro Blue Tetrazolium Chloride, Molecular Biology Reagent Grade, MP Biomedicals™
CAS: 298-83-9 Molecular Formula: C40H30Cl2N10O6 Molecular Weight (g/mol): 817.644 MDL Number: MFCD00012159 InChI Key: FSVCQIDHPKZJSO-UHFFFAOYSA-L Synonym: nitrotetrazolium blue chloride,nitro bt,nitro blue tetrazolium,nbt,nitro blue tetrazolium chloride,p-nitrotetrazolium blue,nitrotetrazolium blue,nitroblue tetrazolium chloride,p-nitro blue tetrazolium chloride,nitro tetrazolium bt PubChem CID: 9281 ChEBI: CHEBI:9505 IUPAC Name: 2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride SMILES: COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-]
Nitro Blue Tetrazolium chloride, 98+%
CAS: 298-83-9 Molecular Formula: C40H30Cl2N10O6 Molecular Weight (g/mol): 817.644 MDL Number: MFCD00012159 InChI Key: FSVCQIDHPKZJSO-UHFFFAOYSA-L Synonym: Nitro BT; Nitrotetrazolium Blue chloride PubChem CID: 9281 ChEBI: CHEBI:9505 IUPAC Name: 2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride SMILES: COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-]
Nitro Blue Tetrazolium 98.0+%, TCI America™
CAS: 298-83-9 Molecular Formula: C40H30Cl2N10O6 Molecular Weight (g/mol): 817.644 MDL Number: MFCD00012159 InChI Key: FSVCQIDHPKZJSO-UHFFFAOYSA-L Synonym: 2,2′C-Di-p-nitrophenyl-5,5′C-diphenyl-(3,3′C-dimethoxy)-4,4′C-bisphenyleneditetrazolium Chloride, NBT PubChem CID: 9281 ChEBI: CHEBI:9505 IUPAC Name: 2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride SMILES: COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-]
Nitro Blue Tetrazolium, Fisher BioReagents
CAS: 298-83-9 Molecular Formula: C40H30Cl2N10O6 Molecular Weight (g/mol): 817.644 InChI Key: FSVCQIDHPKZJSO-UHFFFAOYSA-L Synonym: NBT,Nitro BT,p-Nitro Blue Tetrazolium PubChem CID: 9281 ChEBI: CHEBI:9505 IUPAC Name: 2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride SMILES: COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-]
p-Nitro Blue Tetrazolium Chloride (NBT), Ultrapure, Thermo Scientific™
CAS: 298-83-9 Molecular Formula: C40H30Cl2N10O6 Molecular Weight (g/mol): 817.644 MDL Number: MFCD00012159 InChI Key: FSVCQIDHPKZJSO-UHFFFAOYSA-L Synonym: nitrotetrazolium blue chloride,nitro bt,nitro blue tetrazolium,nbt,nitro blue tetrazolium chloride,p-nitrotetrazolium blue,nitrotetrazolium blue,nitroblue tetrazolium chloride,p-nitro blue tetrazolium chloride,nitro tetrazolium bt PubChem CID: 9281 ChEBI: CHEBI:9505 IUPAC Name: 2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride SMILES: COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-]