Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrobenzene, 99.5%, for analysis

Catalog No. p-4813584
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC447652500 250 mL Glass bottle
AC447650010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC447652500 Supplier Thermo Scientific Chemicals Supplier No. 447652500
Only null left
Add to Cart

CAS: 98-95-3 | C6H5NO2 | 123.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-95-3
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.11
InChI Key LQNUZADURLCDLV-UHFFFAOYSA-N
Synonym nitrobenzol, benzene, nitro, oil of mirbane, mirbane oil, essence of mirbane, nitro-benzene, nitrobenzeen, oil of myrbane, nitrobenzen, mononitrobenzene
PubChem CID 7416
ChEBI CHEBI:27798
IUPAC Name nitrobenzene
SMILES C1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 5.0°C to 6.0°C
Density 1.2050g/mL
Boiling Point 210.0°C to 211.0°C
Flash Point 88°C
Assay Percent Range 99.4% min. (GC)
Packaging Glass bottle
Refractive Index 1.5503 to 1.5523
Linear Formula C6H5NO2
Quantity 250 mL
Merck Index 15,6674
Specific Gravity 1.205
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,benzene,ether and oils
Formula Weight 123.11
Percent Purity 99.5%
Grade Analysis
Chemical Name or Material Nitrobenzene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Toxic in contact with skin.
Harmful to aquatic life with long lasting effects.
Toxic if sw
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove
EINECSNumber 202-716-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.