Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Nitrocefin, Beta-Lactamase Substrate, Solid, 25 mg

Catalog No. 502583249
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3249 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3249 Supplier Abcam Supplier No. AB14562525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chromogenic cephalosporin substrate for the detection of beta-lactamase activity.

  • Molecular weight: 516.5 Da
  • Purity: >95%
  • Form: Solid
  • Application: Chromogenic cephalosporin β-lactamase substrate
  • Detection: Indicates β-lactamase activity via a color change from yellow (max at pH 7.0 = 390 nm) to red (max at pH 7.0 = 486 nm) upon hydrolysis of the β-lactam ring amide bond
  • Storage: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Beta-Lactamase
Substrate Properties Chemical Substrate
Substrate Nitrocefin
Shipping Condition Ambient
Content And Storage Store at -20°C.
Detection Method Chromogenic
Form Solid
Formulation 100% Nitrocefin
Quantity 25 mg
For Use With (Application) Beta-lactamase activity detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.