Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitron 95%, Thermo Scientific™

Catalog No. AC415950250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC415950250 25 g Glass bottle
AC415951000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC415950250 Supplier Thermo Scientific Chemicals Supplier No. 415950250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2218-94-2
Molecular Formula C20H16N4
Molecular Weight (g/mol) 312.376
InChI Key CWGBFIRHYJNILV-UHFFFAOYSA-N
Synonym nitron, unii-90im4z883z, 1,4-diphenyl-endoanilino-dihydrotriazole, 1,4-diphenyl-3-phenylamino-1h-1,2,4-triazolium inner salt, 1h-1,2,4-triazolium, 1,4-diphenyl-3-phenylamino-, hydroxide, inner salt, 1,4-diphenyl-3-phenylazamethylene-1,2,4-triazoline, 1,4-diphenyl-3-phenylamino-1,2,4-triazolium hydroxide inner salt, 4h-1,2,4-triazolium, 1,4-diphenyl-3-phenylamino-, inner salt, 4,5-dihydro-2,4-diphenyl-5-phenylimino-1h-1,2,4-triazolium inner salt
PubChem CID 720071
IUPAC Name (1,4-diphenyl-1,2,4-triazol-4-ium-3-yl)-phenylazanide
SMILES C1=CC=C(C=C1)[N-]C2=NN(C=[N+]2C3=CC=CC=C3)C4=CC=CC=C4

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Brown-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 25 g
Beilstein 26, IV, 1075
Merck Index 15, 6700
Formula Weight 312.36
Physical Form Powder
Chemical Name or Material Nitron, 95%
Show More Show Less
EINECSNumber 218-724-2
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.