Learn More
Sigma Aldrich Fine Chemicals Biosciences Nitroreductase from Escher1MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences N92841MG
Encompass
Nitroreductase is a flavoprotein and is encoded by the NfsB gene. It comprises a dimer with 217 amino acids and active site in each subunit. The structure has fMN and the substrate bound to the enzyme.
Catalog No. 50-290-4535
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.