Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitroterephthalic Acid 98.0+%, TCI America™
SDP

Catalog No. N027225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N027225G 25 g
N02725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N027225G Supplier TCI America Supplier No. N027225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 610-29-7
Molecular Formula C8H5NO6
Molecular Weight (g/mol) 211.129
MDL Number MFCD00007141
InChI Key QUMITRDILMWWBC-UHFFFAOYSA-N
Synonym nitroterephthalic acid, 2-nitroterephthalic acid, 2-nitrobenzene-1,4-dicarboxylic acid, 1,4-benzenedicarboxylic acid, 2-nitro, terephthalic acid, nitro, 2-nitroterephthalicacid, 2-nitroterephthalate, 5-nitroterephthalic acid, acmc-1au2u
PubChem CID 69122
SMILES C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])C(=O)O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 211.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nitroterephthalic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.