Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Includes
- Streptavidin Capture Plate: One pre-coated 96-stripwell immunoplate sealed in a foil pouch
- U-Bottom Plate: One clear non-binding U-bottom plate
- TBS, 20X
- 20% Tween™ 20
- LumiGlo Chemiluminescent Substrate Reagent A
- LumiGlo Chemiluminescent Substrate Reagent B
- Plate Covers: Two plate covers
- Nitrotyrosine Peptide Standard
- 1000X Biotinylated Nitrotyrosine Peptide
- 2000X Anti-Nitrotyrosine Detection Antibody
- Gt X Rb, HRP Secondary Antibody
- 30% BSA
Requires
- Multi-channel or repeating pipettes
- 96-well microtiter plate reader with luminescence readout.
- Mechanical vortex
- The kit includes all reagents needed for measurement of tyrosine nitration, including a pre-coated 96-well plates, Nitrotyrosine peptide standard, a Nitrotyrosine antibody, LumiGLO™ chemiluminescent detection substrate, and wash buffers
- The assay has a wide dynamic range and high precision, making this assay a valuable new tool for the study of tyrosine nitration
- Application: This Nitrotyrosine ELISA is used to measure & quantify Nitrotyrosine levels in Neuroscience and Toxicity research
Specifications
Specifications
| Content And Storage | Avoid repeated freeze/thaw cycles, aliquot if necessary |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?