Learn More
Nitrovin, G-Biosciences

Description
This reagent is a nitrofuran and guanidine which can be applied in chemistry as an organic building block.
Specifications
Specifications
| Quantity | 100 mg |
| Boiling Point | 492.33°C |
| Product Line | Gibco |
| Physical Form | Powder |
| Purity | 98% |
| Molecular Weight (g/mol) | 360.28 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.