Learn More
Lonza Walkersville NK - Human Natural Killer Cells, Cryopreserved, Negative Selection, 5 Million
Supplier:  Lonza Walkersville 2W501
Encompass
Natural Killer (NK) cells are lymphocytes of the immune system that are critical in host defense and immune regulation as part of both the innate and adaptive immune response. They are of particular interest to immunotherapy researchers as they hold the promise to specifically target and destroy cancer cells and tumors while leaving healthy cells intact.Cells You Can Count On:Natural Killer cells are traditionally characterized by the presence of the CD56 cell surface marker and absence of CD3. In addition, CD16 is important for NK cell function because CD16 induces cytokine expression and cytotoxic effector activity.Benefits:Test negative for mycoplasma, bacteria, yeast, and fungiHIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lotsA Certificate ofAnalysis is provided for each cell lot purchased Content:1 x Cryopreserved ampule of Human Peripheral Blood NK containing ≥ 5 million cells
Catalog No. NC1961068
Only null left
Add to cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.