Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NKAIN4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556280
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556280 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556280 Supplier Invitrogen™ Supplier No. PA556280
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LKDSELLTFS LSRHRSWWRE RWPGCLHEEV PAVGLGAPHG QALVSGAGCA LEPSYVEALH SCLQILIALL GFVCGCQVVS VFTEEEDSFD FIGGFDPFPL YHVNEKPSSL LSKQVY Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 77%.

NKAIN4 is a member of a family of mammalian proteins (see NKAIN1; MIM 612871) with similarity to Drosophila Nkain and interacts with the beta subunit of Na,K-ATPase (ATP1B1; MIM 182330) (Gorokhova et al., 2007 [PubMed 17606467]).[supplied by OMIM, Jun 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKAIN4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NKAIN4
Gene Accession No. Q8IVV8
Gene Alias bA261N11.2; C20orf58; FAM77A; Na(+)/K(+)-transporting ATPase subunit beta-1-interacting protein 4; Na+/K+ transporting ATPase interacting 4; NKAIN4; Protein FAM77A; Sodium/potassium-transporting ATPase subunit beta-1-interacting protein 4
Gene Symbols NKAIN4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NKAIN4. Recombinant protein control fragment (Product #RP-90926).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128414
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.