Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NKCC1 Monoclonal Antibody (6G7D2)
GREENER_CHOICE

Catalog No. PIMA549260 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549260 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549260 Supplier Invitrogen™ Supplier No. MA549260
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human A549 whole cell, human CACO-2 whole cell, human PC-3 whole cell. IHC: human colorectal adenocarcinoma tissue, mouse colon tissue, rat colon tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

NKCC1 mediates sodium and chloride transport and reabsorption. NKCC1 is a membrane protein and is important in maintaining proper ionic balance and cell volume. NKCC1 is an electrically silent transporter system that mediates sodium and chloride reabsorption. Further, NKCC1 plays a vital role in the regulation of ionic balance and cell volume and is phosphorylated in response to DNA damage. Three transcript variants encoding two different isoforms have been found for NKCC1. Diseases associated with SLC12A2 include Basosquamous Carcinoma and Metatypical Basal Cell Carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKCC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 6G7D2
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene SLC12A2
Gene Accession No. P55011, P55012
Gene Alias 9330166H04Rik; basolateral NaK(2Cl) cotransporter; basolateral Na-K-Cl symporter; BSC; BSC2; bumetanide-sensitive sodium-(potassium)-chloride cotransporter 1; Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 2; little ears; lte; mBSC2; Na-K-2Cl- cotransporter; Na-K-Cl cotransporter; NKCC1; PPP1R141; protein phosphatase 1, regulatory subunit 141; Slc12a2; slc12a4; sodium/potassium/chloride cotransporters; sodium/potassium/chloride transporters; solute carrier family 12 (potassium/chloride transporters), member 2; solute carrier family 12 (potassium/chloride transporters), member 4; solute carrier family 12 (sodium/potassium/chloride transporter), member 2; solute carrier family 12 (sodium/potassium/chloride transporters), member 2; solute carrier family 12 member 2; solute carrier family 12, member 2; sy-ns; zfnkcc1; zgc:85961; zmp:0000000537
Gene Symbols SLC12A2
Host Species Mouse
Immunogen E.coli-derived human NKCC1/SLC12A2 recombinant protein (Position: K889-K943).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20496, 6558, 83629
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.