Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKG2A/CD159a/KLRC1 Antibody (131411R) [Janelia Fluor™ 525], R&D Systems™
SDP

Catalog No. NB474002
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB474002 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB474002 Supplier R&D Systems Supplier No. FAB1059RJF525

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

NKG2A/CD159a/KLRC1 Monoclonal specifically detects NKG2A/CD159a/KLRC1 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen NKG2A/CD159a/KLRC1
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 131411R
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias CD159 antigen-like family member A, CD159A, CD159a antigen, C-lectin type II protein, killer cell lectin-like receptor subfamily C, member 1, MGC13374, natural killer cell lectin, natural killer group protein 2, NK cell receptor A, NKG2, NKG2-1/B activating NK receptor, NKG2-A, NKG2-A/B type II integral membrane protein, NKG2-A/B-activating NK receptor, NKG2-A/NKG2-B type II integral membrane protein, NKG2AMGC59791, NKG2-B
Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human NKG2A and CD94
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3821
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Liquid
Isotype IgG2a
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.