Learn More
Sino Biological Mouse NKG2D / CD314 Protein (Fc Tag) 100ug
Supplier:  Sino Biological 57340M01B100
Encompass_Preferred
A DNA sequence encoding the mouse KLRK1 (NP_001076791.1) (Phe77-Val219) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-211-4930
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.